4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}aniline
Lieferant: Apollo Scientific
|
Gefahr
|
PC6080-1G
APOSPC6080-1GEA
113
EUR
APOSPC6080-1G
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}aniline
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}aniline
Formel:
C₁₂H₉F₃N₂O Molecular Weight: 254.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10703507 CAS-Nummer: 1086378-51-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...