3,5-Dichloro-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}aniline

Lieferant: Apollo Scientific

Synonyme: 2-(4-Amino-2,6-dichlorophenoxy)-6-(trifluoromethyl)pyridine

PC6081-1G
APOSPC6081-1GEA 241 EUR
APOSPC6081-1G
3,5-Dichloro-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}aniline
3,5-Dichloro-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}aniline
Formel: C₁₂H₇Cl₂F₃N₂O
Molecular Weight: 323,098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109517
CAS-Nummer: 1211176-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR