1,1,1,2,3,3,3-Heptafluor-2-(trifluormethyl)propan 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Perfluoroisobutane

PC6088W-5G
APOSPC6088W-5GEA 628 EUR
APOSPC6088W-5G
1,1,1,2,3,3,3-Heptafluor-2-(trifluormethyl)propan 95%
1,1,1,2,3,3,3-Heptafluor-2-(trifluormethyl)propan

Contains up to 5% perfluorobutane

Formel: C₄F₁₀
Siedepunkt: 0 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 354-92-7
UN: 3163
ADR: 2.2,

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR