1-Iod-5-(trifluormethyl)perfluorhexan 97%
Lieferant: Apollo Scientific
PC6089-25G
APOSPC6089-25GEA
698
EUR
APOSPC6089-25G
1-Iod-5-(trifluormethyl)perfluorhexan 97%
1-Iod-5-(trifluormethyl)perfluorhexan
Formel:
C₇F₁₅I Molecular Weight: 495,96 g/mol Siedepunkt: 139 °C Dichte: 2,039 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042357 CAS-Nummer: 3486-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...