2-Phenoxy-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC6091-1G
APOSPC6091-1GEA 147 EUR
APOSPC6091-1G
2-Phenoxy-6-(trifluoromethyl)pyridine
2-Phenoxy-6-(trifluoromethyl)pyridine
Formel: C₁₂H₈F₃NO
Molecular Weight: 239,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109518
CAS-Nummer: 1209243-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR