4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride

Lieferant: Apollo Scientific

Synonyme: 4-(6-Trifluoromethyl-2-pyridyloxy)benzenesulphonyl chloride

PC6092-1G
APOSPC6092-1GEA 241 EUR
APOSPC6092-1G
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride
Formel: C₁₂H₇ClF₃NO₃S
Molecular Weight: 337,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100129
CAS-Nummer: 1210732-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR