1,1'-Difluor-2,2'-bipyridiniumditetrafluoroborat 98%

Lieferant: Apollo Scientific
Gefahr

PC6095-25G PC6095-5G
APOSPC6095-25GEA 1220 EUR
APOSPC6095-25G APOSPC6095-5G
1,1'-Difluor-2,2'-bipyridiniumditetrafluoroborat 98%
1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate)

Fluorinating agent (103g/Kg effective F2), ask for literature

Formel: C₁₀H₈B₂F₁₀N₂
Schmelzpunkt: 166…175 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD01073546
CAS-Nummer: 178439-26-4
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR