4-Amino-3-fluorphenol Hydrochlorid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC6096-25G
APOSPC6096-25GEA
102
EUR
APOSPC6096-25G
4-Amino-3-fluorphenol Hydrochlorid 95%
4-Amino-3-fluorphenol Hydrochlorid
Formel:
C₆H₇ClFNO Molecular Weight: 163.5773 g/mol Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01632189 CAS-Nummer: 18266-53-0 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...