Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 5-iodo-3-methyl-2-[(trifluoroacetyl)amino]benzoate

PC6102-1G PC6102-500MG PC6102-5G
APOSPC6102-1GEA 161 EUR
APOSPC6102-1G APOSPC6102-500MG APOSPC6102-5G
Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate
Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate
Formel: C₁₁H₉F₃INO₃
Molecular Weight: 387.1 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12827837
CAS-Nummer: 1186405-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR