Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-iodo-3-methyl-2-[(trifluoroacetyl)amino]benzoate
PC6102-1G
PC6102-500MG
PC6102-5G
APOSPC6102-1GEA
161
EUR
APOSPC6102-1G
APOSPC6102-500MG
APOSPC6102-5G
Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate
Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate
Formel:
C₁₁H₉F₃INO₃ Molecular Weight: 387.1 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12827837 CAS-Nummer: 1186405-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...