2-(4-Amino-3-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol

Lieferant: Apollo Scientific

Synonyme: 4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)-2-methylaniline, 2-Amino-5-(1,1,1,3,3,3-hexafluoro-2-hydroxyprop-2-yl)toluene

PC6106-1G
APOSPC6106-1GEA 194 EUR
APOSPC6106-1G
2-(4-Amino-3-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
2-(4-Amino-3-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Formel: C₁₀H₉F₆NO
Molecular Weight: 273,175 g/mol
Schmelzpunkt: 135…136 °C
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00788428
CAS-Nummer: 1992-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR