2-(Methylsulphonyl)-5-nitrobenzotrifluoride 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Methylsulphonyl)-4-nitro-2-(trifluoromethyl)benzene, Methyl 4-nitro-2-(trifluoromethyl)phenyl sulphone

PC6112-1G
APOSPC6112-1GEA 125 EUR
APOSPC6112-1G
2-(Methylsulphonyl)-5-nitrobenzotrifluoride 99%
2-(Methylsulphonyl)-5-nitrobenzotrifluoride
Formel: C₈H₆F₃NO₄S
Molecular Weight: 269,198 g/mol
Schmelzpunkt: 131…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13176597
CAS-Nummer: 252561-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR