2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Formyl-2-methyl-4-(trifluoromethyl)-1,3-thiazole
PC6115-1G
APOSPC6115-1GEA
389
EUR
APOSPC6115-1G
2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxaldehyde 97%
2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxaldehyde
Formel:
C₆H₄F₃NOS Molecular Weight: 195,162 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12198128 CAS-Nummer: 1034566-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...