2-(Perfluoro-7-methyloctyl)ethyl iodide 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H,1H,2H,2H-Hexadecafluoro-9-(trifluoromethyl)decyl iodide
PC6120H-250MG
APOSPC6120H-250MGEA
14.7
EUR
APOSPC6120H-250MG
2-(Perfluoro-7-methyloctyl)ethyl iodide 98%
2-(Perfluoro-7-methyloctyl)ethyl iodide
Formel:
C₁₁H₄F₁₉I Molecular Weight: 624,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153247 CAS-Nummer: 40678-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...