2-(Perfluoro-5-methylhexyl)ethyl methacrylate 97%

Lieferant: Apollo Scientific
Achtung

PC6121M-5G
APOSPC6121M-5GEA 0 EUR
APOSPC6121M-5G
2-(Perfluoro-5-methylhexyl)ethyl methacrylate 97%
2-(Perfluoro-5-methylhexyl)ethyl methacrylate

[1H,1H,2H,2H-Perfluoro(7-methyloctyl) methacrylate]n 1.3470
d 1.567

Formel: C₁₃H₉F₁₅O₂
Dichte: 1,567 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00236098
CAS-Nummer: 50836-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR