Methyl perfluoro(2-methyl-3-oxahexanoate) 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-(heptafluoropropoxy)-2,3,3,3-tetrafluoropropanoate, Methyl undecafluoro-2-methyl-3-oxahexanoate, Methyl 2-(heptafluoropropoxy)-2,3,3,3-tetrafluoropropionate
PC6122D-1G
APOSPC6122D-1GEA
46.4
EUR
APOSPC6122D-1G
Methyl perfluoro(2-methyl-3-oxahexanoate) 97%
Methyl perfluoro(2-methyl-3-oxahexanoate)
Formel:
C₇H₃F₁₁O₃ Molecular Weight: 344,079 g/mol Siedepunkt: 115 - 117 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00042358 CAS-Nummer: 13140-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...