4-Fluoroisophthalonitrile 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluorobenzene-1,3-dicarbonitrile
PC6129-1G
APOSPC6129-1GEA
79.9
EUR
APOSPC6129-1G
4-Fluoroisophthalonitrile 99%
4-Fluoroisophthalonitrile
Formel:
C₈H₃FN₂ Molecular Weight: 146.121 g/mol Schmelzpunkt: 89…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01657003 CAS-Nummer: 13519-90-9 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...