3-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC6142-1G
PC6142-250MG
APOSPC6142-1GEA
252
EUR
APOSPC6142-1G
APOSPC6142-250MG
3-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
3-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Formel:
C₈H₄BrF₃N₂ Schmelzpunkt: 98…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111688 CAS-Nummer: 503172-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...