2-Chloro-3,6-difluoro-4-hydroxybenzenecarboxylic acid

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-3,6-difluoro-4-hydroxybenzoic acid

PC6148-500MG
APOSPC6148-500MGEA 473 EUR
APOSPC6148-500MG
2-Chloro-3,6-difluoro-4-hydroxybenzenecarboxylic acid
2-Chloro-3,6-difluoro-4-hydroxybenzenecarboxylic acid
Formel: C₇H₃ClF₂O₃
Molecular Weight: 208,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025951
CAS-Nummer: 749230-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR