4,5-Dibromo-1-methyl-1H-imidazole-2-carbaldehyde

Lieferant: Apollo Scientific
Achtung

PC6152-500MG
APOSPC6152-500MGEA 236 EUR
APOSPC6152-500MG
4,5-Dibromo-1-methyl-1H-imidazole-2-carbaldehyde
4,5-Dibromo-1-methyl-1H-imidazole-2-carbaldehyde
Formel: C₅H₄Br₂N₂O
Molecular Weight: 267,92 g/mol
Schmelzpunkt: 167…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12025924
CAS-Nummer: 79326-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR