2,6-Dichloro-4-(trifluoromethyl)nicotinic acid

Lieferant: Apollo Scientific
Achtung

PC6153-1G PC6153-500MG
APOSPC6153-1GEA 295 EUR
APOSPC6153-1G APOSPC6153-500MG
2,6-Dichloro-4-(trifluoromethyl)nicotinic acid
2,6-Dichloro-4-(trifluoromethyl)nicotinic acid
Formel: C₇H₂Cl₂F₃NO₂
Schmelzpunkt: 79…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06208180
CAS-Nummer: 503437-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR