Methyl-2,6-dichloro-4-(trifluoromethyl)nicotinate

Lieferant: Apollo Scientific

Synonyme: Methyl 2,6-dichloro-4-(trifluoromethyl)nicotinate

PC6157-500MG
APOSPC6157-500MGEA 547 EUR
APOSPC6157-500MG
Methyl-2,6-dichloro-4-(trifluoromethyl)nicotinate
Methyl-2,6-dichloro-4-(trifluoromethyl)nicotinate
Formel: C₈H₄Cl₂F₃NO₂
Molecular Weight: 274.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025929
CAS-Nummer: 1130344-76-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR