2-Piperazin-1-yl-4-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyrimidine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-Cyclopropyl-1,2,4-oxadiazol-3-yl)-2-(piperazin-1-yl)pyrimidine dihydrochloride

PC6158-500MG
APOSPC6158-500MGEA 684 EUR
APOSPC6158-500MG
2-Piperazin-1-yl-4-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyrimidine dihydrochloride
2-Piperazin-1-yl-4-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyrimidine dihydrochloride
Formel: C₁₁H₁₃Cl₂F₃N₆O
Molecular Weight: 373,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025837
CAS-Nummer: 1177093-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR