2-(Trifluoromethyl)imidazo[1,2-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC6159-1G
PC6159-500MG
APOSPC6159-1GEA
270
EUR
APOSPC6159-1G
APOSPC6159-500MG
2-(Trifluoromethyl)imidazo[1,2-a]pyridine
2-(Trifluoromethyl)imidazo[1,2-a]pyridine
Formel:
C₈H₅F₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111687 CAS-Nummer: 73221-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...