8H-Perfluorooctyl iodide

Lieferant: Apollo Scientific
Achtung

PC6171B-5G
APOSPC6171B-5GEA 78.6 EUR
APOSPC6171B-5G
8H-Perfluorooctyl iodide
8H-Perfluorooctyl iodide
Formel: C₈HF₁₆I
Lagertemperatur: Raumtemperatur
MDL: MFCD00236712
CAS-Nummer: 79162-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR