Nonafluorovaleric acid
Lieferant: Apollo Scientific
|
Gefahr
|
PC6184-25G
PC6184-5G
APOSPC6184-25GEA
0
EUR
APOSPC6184-25G
APOSPC6184-5G
Nonafluorovaleric acid
Nonafluorovaleric acid
Formel:
C₅HF₉O₂ Siedepunkt: 82 °C/85mmHg Dichte: 1,713 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040211 CAS-Nummer: 2706-90-3 EINECS: 220-300-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...