4-(Trifluoroacetyl)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Formyl-2,2,2-trifluoroacetophenone, 4-(2,2,2-trifluoroacetyl)benzaldehyde, 1-(4-Formylphenyl)-2,2,2-trifluoroethan-1-one
PC6194-1G
PC6194-250MG
PC6194-5G
APOSPC6194-1GEA
263
EUR
APOSPC6194-1G
APOSPC6194-250MG
APOSPC6194-5G
4-(Trifluoroacetyl)benzaldehyde 95%
4-(Trifluoroacetyl)benzaldehyde
Formel:
C₉H₅F₃O₂ Molecular Weight: 202.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12547943 CAS-Nummer: 86988-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...