4-(Trifluoroacetyl)benzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Formyl-2,2,2-trifluoroacetophenone, 4-(2,2,2-trifluoroacetyl)benzaldehyde, 1-(4-Formylphenyl)-2,2,2-trifluoroethan-1-one

PC6194-1G PC6194-250MG PC6194-5G
APOSPC6194-1GEA 263 EUR
APOSPC6194-1G APOSPC6194-250MG APOSPC6194-5G
4-(Trifluoroacetyl)benzaldehyde 95%
4-(Trifluoroacetyl)benzaldehyde
Formel: C₉H₅F₃O₂
Molecular Weight: 202.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12547943
CAS-Nummer: 86988-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR