4'-(Trifluoroacetyl)acetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Acetylphenyl)-2,2,2-trifluoroethan-1-one, 1-(4-Acetylphenyl)-2,2,2-trifluoroethanone

PC6195-1G PC6195-250MG PC6195-5G
APOSPC6195-1GEA 192 EUR
APOSPC6195-1G APOSPC6195-250MG APOSPC6195-5G
4'-(Trifluoroacetyl)acetophenone
4'-(Trifluoroacetyl)acetophenone
Formel: C₁₀H₇F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12547944
CAS-Nummer: 86988-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR