2-Brom-4-fluor-5-(trifluormethyl)anilin

Lieferant: Apollo Scientific

Synonyme: 6-Brom-α,α,α,4-tetrafluor-m-toluidin

PC6197-10G PC6197-5G PC6197-1G
APOSPC6197-10GEA 192 EUR
APOSPC6197-10G APOSPC6197-5G APOSPC6197-1G
2-Brom-4-fluor-5-(trifluormethyl)anilin
2-Brom-4-fluor-5-(trifluormethyl)anilin
Formel: C₇H₄BrF₄N
Molecular Weight: 258.01 g/mol
Schmelzpunkt: 24…29 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04973755
CAS-Nummer: 193090-60-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR