4-Brom-3-chlorbenzotrifluorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-chloro-4-trifluoromethylbenzene
PC6198-100G
APOSPC6198-100GEA
137
EUR
APOSPC6198-100G
4-Brom-3-chlorbenzotrifluorid 98%
4-Brom-3-chlorbenzotrifluorid
Formel:
C₇H₃BrClF₃ Molecular Weight: 259,45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00673985 CAS-Nummer: 402-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...