Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetra-oxapentadecanoic acid, methyl ester 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC6219D-25G
PC6219D-5G
PC6219D-1G
APOSPC6219D-25GEA
551
EUR
APOSPC6219D-25G
APOSPC6219D-5G
APOSPC6219D-1G
Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetra-oxapentadecanoic acid, methyl ester 97%
Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetra-oxapentadecanoic acid, methyl ester
Formel:
C₁₆H₃F₂₉O₆ Siedepunkt: 70…80 °C/0,1mmHg Dichte: 1,74 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054670 CAS-Nummer: 133609-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...