Toluol-α-sulfonylfluorid 99%

Lieferant: Apollo Scientific
Gefahr

PC6222M-100G PC6222M-250G PC6222M-25G
APOSPC6222M-100GEA 149 EUR
APOSPC6222M-100G APOSPC6222M-250G APOSPC6222M-25G
Toluol-α-sulfonylfluorid 99%
Toluol-α-sulfonylfluorid

Inhibitor of Serine proteases: Toxicity LD50 <500mg/kgAffords alkenes via reaction with carbonyl compounds

Formel: C₇H₇FO₂S
Schmelzpunkt: 92…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007424
CAS-Nummer: 329-98-6
EINECS: 206-350-2
UN: 2928
ADR: 6.1+8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR