5-Fluorobenzo[b]furan-3(2H)-one 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-5-fluoro-3-oxobenzo[b]furan, 5-Fluorobenzofuran-3(2H)-one, 5-Fluoro-1-benzofuran-3(2H)-one

PC6228-1G
APOSPC6228-1GEA 132 EUR
APOSPC6228-1G
5-Fluorobenzo[b]furan-3(2H)-one 95%
5-Fluorobenzo[b]furan-3(2H)-one
Formel: C₈H₅FO₂
Molecular Weight: 152,123 g/mol
Schmelzpunkt: 102…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08544409
CAS-Nummer: 60770-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR