6-Brom-5,8-difluorchinolin

Lieferant: Apollo Scientific

PC6235-1G
APOSPC6235-1GEA 524 EUR
APOSPC6235-1G
6-Brom-5,8-difluorchinolin
6-Bromo-5,8-difluoroquinoline
Formel: C₉H₄BrF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD11855886
CAS-Nummer: 1133115-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR