7-Chloro-8-fluoroquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Chloro-8-fluoro-1-azanaphthalene

PC6236-1G
APOSPC6236-1GEA 350 EUR
APOSPC6236-1G
7-Chloro-8-fluoroquinoline
7-Chloro-8-fluoroquinoline
Formel: C₉H₅ClFN
Molecular Weight: 181,594 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11855931
CAS-Nummer: 1133116-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR