6-Fluoro-2-naphthonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Cyano-6-fluoronaphthalene, 6-Fluoronaphthalene-2-carbonitrile
PC6244-1G
APOSPC6244-1GEA
389
EUR
APOSPC6244-1G
6-Fluoro-2-naphthonitrile
6-Fluoro-2-naphthonitrile
Formel:
C₁₁H₆FN Molecular Weight: 171.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12922839 CAS-Nummer: 13790-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...