3'-Bromo-4'-(trifluoromethoxy)acetanilide 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC6257-1G
PC6257-5G
APOSPC6257-1GEA
55.1
EUR
APOSPC6257-1G
APOSPC6257-5G
3'-Bromo-4'-(trifluoromethoxy)acetanilide 99%
3'-Bromo-4'-(trifluoromethoxy)acetanilide
Formel:
C₉H₇BrF₃NO₂ Schmelzpunkt: 86…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13176598 CAS-Nummer: 755030-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...