Poly(vinylidene fluoride)

Lieferant: Apollo Scientific
Achtung

PC6267-25G PC6267-100G PC6267-250G
APOSPC6267-25GEA 21.9 EUR
APOSPC6267-25G APOSPC6267-100G APOSPC6267-250G
Poly(vinylidene fluoride)
Poly(vinylidene fluoride)
Formel: (-CH₂CF₂-)n
Schmelzpunkt: 155…165 °C
Dichte: 1.74 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00084470
CAS-Nummer: 24937-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR