Poly(vinylidene fluoride)
Lieferant: Apollo Scientific
|
Achtung
|
PC6267-25G
PC6267-100G
PC6267-250G
APOSPC6267-25GEA
21.9
EUR
APOSPC6267-25G
APOSPC6267-100G
APOSPC6267-250G
Poly(vinylidene fluoride)
Poly(vinylidene fluoride)
Formel:
(-CH₂CF₂-)n Schmelzpunkt: 155…165 °C Dichte: 1.74 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084470 CAS-Nummer: 24937-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...