Poly-4-vinylpyridinium poly(hydrogen fluoride)

Lieferant: Apollo Scientific
Gefahr

PC6267G-25G
APOSPC6267G-25GEA 394 EUR
APOSPC6267G-25G
Poly-4-vinylpyridinium poly(hydrogen fluoride)
Poly-4-vinylpyridinium poly(hydrogen fluoride)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 79821-63-9
UN: 3261
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR