3,4-Difluorostyrene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Difluoro-4-ethenylbenzene, 1,2-Difluoro-4-vinylbenzene
PC6276-1G
PC6276-25G
PC6276-5G
APOSPC6276-1GEA
37.7
EUR
APOSPC6276-1G
APOSPC6276-25G
APOSPC6276-5G
3,4-Difluorostyrene
3,4-Difluorostyrene
Formel:
C₈H₆F₂ Molecular Weight: 140.13 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09038471 CAS-Nummer: 405-03-8 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...