3,4-Difluorostyrene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Difluoro-4-ethenylbenzene, 1,2-Difluoro-4-vinylbenzene

PC6276-1G PC6276-25G PC6276-5G
APOSPC6276-1GEA 37.7 EUR
APOSPC6276-1G APOSPC6276-25G APOSPC6276-5G
3,4-Difluorostyrene
3,4-Difluorostyrene
Formel: C₈H₆F₂
Molecular Weight: 140.13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09038471
CAS-Nummer: 405-03-8
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR