3-Oxo-N-(3-(trifluoromethyl)phenyl)butanamide

Lieferant: Apollo Scientific

PC6299-1G
APOSPC6299-1GEA 154 EUR
APOSPC6299-1G
3-Oxo-N-(3-(trifluoromethyl)phenyl)butanamide
3-Oxo-N-(3-(trifluoromethyl)phenyl)butanamide
Formel: C₁₁H₁₀F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00201876
CAS-Nummer: 785-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR