2-(Trifluoromethyl)xanthone

Lieferant: Apollo Scientific

PC6302-1G
APOSPC6302-1GEA 245 EUR
APOSPC6302-1G
2-(Trifluoromethyl)xanthone
2-(Trifluoromethyl)xanthone
Formel: C₁₄H₇F₃O₂
Molecular Weight: 264,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701090
CAS-Nummer: 1496-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR