1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazol-4-amine
Lieferant: Apollo Scientific
PC6304-1G
PC6304-10G
PC6304-5G
APOSPC6304-1GEA
217
EUR
APOSPC6304-1G
APOSPC6304-10G
APOSPC6304-5G
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazol-4-amine
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazol-4-amine
Formel:
C₈H₆F₃N₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505006 CAS-Nummer: 1210732-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...