2-Fluoro-5-iodoaniline
Lieferant: Apollo Scientific
|
Achtung
|
PC6306-5G
APOSPC6306-5GEA
68.3
EUR
APOSPC6306-5G
2-Fluoro-5-iodoaniline
2-Fluoro-5-iodoaniline
Formel:
C₆H₅FIN Lagertemperatur: Raumtemperatur |
MDL:
MFCD06796596 CAS-Nummer: 886362-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...