2-Fluoro-3-(trifluoromethoxy)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC6313-1G PC6313-5G PC6313-25G
APOSPC6313-1GEA 0 EUR
APOSPC6313-1G APOSPC6313-5G APOSPC6313-25G
2-Fluoro-3-(trifluoromethoxy)benzyl bromide 95%
2-Fluoro-3-(trifluoromethoxy)benzyl bromide
Formel: C₈H₅BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD12026474
CAS-Nummer: 1159512-59-0
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR