2-Fluoro-3-(trifluoromethoxy)benzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC6313-1G
PC6313-5G
PC6313-25G
APOSPC6313-1GEA
0
EUR
APOSPC6313-1G
APOSPC6313-5G
APOSPC6313-25G
2-Fluoro-3-(trifluoromethoxy)benzyl bromide 95%
2-Fluoro-3-(trifluoromethoxy)benzyl bromide
Formel:
C₈H₅BrF₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026474 CAS-Nummer: 1159512-59-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...