1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine

Lieferant: Apollo Scientific
Gefahr

PC6316-1G
APOSPC6316-1GEA 289 EUR
APOSPC6316-1G
1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine
1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine
Formel: C₁₄H₁₃Cl₂F₃N₄S
Molecular Weight: 397.25 g/mol
Schmelzpunkt: 97…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172020
CAS-Nummer: 303150-21-2
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR